Skip to Content
MilliporeSigma

BDPH9BCDADA0

Bide Pharmatech

6-(2,5-Dichlorophenyl)-1,3,5-triazine-2,4-diamine

Synonym(s):

6-(2,5-dichlorophenyl)-1,3,5-triazine-2,4-diamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H7N5Cl2
CAS Number:
Molecular Weight:
256.09
MDL number:


Inchi Code

1S/C9H7Cl2N5/c10-4-1-2-6(11)5(3-4)7-14-8(12)16-9(13)15-7/h1-3H,(H4,12,13,14,15,16)

Inchi Key

ATCGGEJZONJOCL-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

99%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

White to off-white powder or crystals



Still not finding the right product?


hstatements

pcodes

signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library