Skip to Content
MilliporeSigma

BDPH9BCDAE33

Bide Pharmatech

Ethyl 3-(5-methylthiophen-2-yl)-3-oxopropanoate

Synonym(s):

ethyl 3-(5-methyl-2-thienyl)-3-oxopropanoate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H12O3S1
CAS Number:
Molecular Weight:
212.27
MDL number:


Inchi Code

1S/C10H12O3S/c1-3-13-10(12)6-8(11)9-5-4-7(2)14-9/h4-5H,3,6H2,1-2H3

Inchi Key

GJZZCAURPLNHGC-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library