Skip to Content
MilliporeSigma

BDPH9BCDB0F0

Bide Pharmatech

4-Chloro-6-iodoquinazoline

Synonym(s):

4-chloro-6-iodoquinazoline

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H4N2Cl1I1
CAS Number:
Molecular Weight:
290.49
MDL number:


Inchi Code

1S/C8H4ClIN2/c9-8-6-3-5(10)1-2-7(6)11-4-12-8/h1-4H

Inchi Key

BDAIUOPDSRAOKI-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Country of Origin

CN

Physical Form

Form: Crystal - Powder / Colour: White - Yellow - Pale-brown

Storage temp.

Keep in dark place, inert atmosphere, 2-8°C



Still not finding the right product?

Explore all of our products under 4-Chloro-6-iodoquinazoline


signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library