Skip to Content
MilliporeSigma

BDPH9BCDB355

Bide Pharmatech

6-Bromophthalazine-1,4-diol

Synonym(s):

6-bromophthalazine-1,4-diol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H5O2N2Br1
CAS Number:
Molecular Weight:
241.04
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H5BrN2O2/c9-4-1-2-5-6(3-4)8(13)11-10-7(5)12/h1-3H,(H,10,12)(H,11,13)

Inchi Key

UFWQSROUAIPXQJ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

White to off-white solid



Still not finding the right product?

Explore all of our products under 6-Bromophthalazine-1,4-diol


hstatements

Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library