Skip to Content
MilliporeSigma

BDPH9BCDB59C

Bide Pharmatech

4-(Morpholinomethyl)phenylboronic acid

Synonym(s):

4-(4-morpholinylmethyl)phenylboronic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C11H16O3N1B1
CAS Number:
Molecular Weight:
221.06
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C11H16BNO3/c14-12(15)11-3-1-10(2-4-11)9-13-5-7-16-8-6-13/h1-4,14-15H,5-9H2

Inchi Key

QPFDUULIDNELSE-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Keep in dark place, sealed in dry, room temperature

Country of Origin

CN

Physical Form

Form: Crystal - Powder - Particles / Colour: White - Slightly pale reddish yellow - Yellow



Still not finding the right product?


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library