Skip to Content
MilliporeSigma

BDPH9BCDB863

Bide Pharmatech

3-(2-Bromophenoxy)propanoic acid

Synonym(s):

3-(2-bromophenoxy)propanoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H9O3Br1
CAS Number:
Molecular Weight:
245.07
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H9BrO3/c10-7-3-1-2-4-8(7)13-6-5-9(11)12/h1-4H,5-6H2,(H,11,12)

Inchi Key

YRCVMOJOHSPJGU-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under 3-(2-Bromophenoxy)propanoic acid



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library