Skip to Content
MilliporeSigma

BDPH9BCDBE03

Bide Pharmatech

1-Bromo-8-(bromomethyl)naphthalene

Synonym(s):

1-bromo-8-(bromomethyl)naphthalene

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C11H8Br2
CAS Number:
Molecular Weight:
299.99
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C11H8Br2/c12-7-9-5-1-3-8-4-2-6-10(13)11(8)9/h1-6H,7H2

Inchi Key

RUGOMNXLAWWDCF-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Powder or crystals



Still not finding the right product?

Explore all of our products under 1-Bromo-8-(bromomethyl)naphthalene



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library