Skip to Content
MilliporeSigma

Skip To

BDPH9BCDC00D

Bide Pharmatech

2-(3-Fluoro-4-methoxyphenyl)ethanol

Synonym(s):

2-(3-fluoro-4-methoxyphenyl)ethanol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
C9H11O2F1
Molecular Weight:
170.18
MDL number:
Pricing and availability is not currently available. Contact Technical Service
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H11FO2/c1-12-9-3-2-7(4-5-11)6-8(9)10/h2-3,6,11H,4-5H2,1H3

Inchi Key

CTZFMMKZSPJHOS-UHFFFAOYSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library