Skip to Content
MilliporeSigma

BDPH9BCDC30E

Bide Pharmatech

Methyl 6-chloro-4-methoxy-1H-pyrrolo[3,2-c]pyridine-2-carboxylate

Synonym(s):

methyl 6-chloro-4-methoxy-1H-pyrrolo[3,2-c]pyridine-2-carboxylate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H9O3N2Cl1
CAS Number:
Molecular Weight:
240.65
MDL number:


Inchi Code

1S/C10H9ClN2O3/c1-15-9-5-3-7(10(14)16-2)12-6(5)4-8(11)13-9/h3-4,12H,1-2H3

Inchi Key

OUFHVWNPQDQNCK-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Country of Origin

CN

Physical Form

Solid

Storage temp.

Inert atmosphere, 2-8°C



Still not finding the right product?


signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library