Skip to Content
MilliporeSigma

BDPH9BCDC313

Bide Pharmatech

1-(tert-Butoxycarbonyl)-3-(trifluoromethyl)piperidine-4-carboxylic acid

Synonym(s):

1-(tert-butoxycarbonyl)-3-(trifluoromethyl)-4-piperidinecarboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C12H18O4N1F3
CAS Number:
Molecular Weight:
297.27
MDL number:


Inchi Code

1S/C12H18F3NO4/c1-11(2,3)20-10(19)16-5-4-7(9(17)18)8(6-16)12(13,14)15/h7-8H,4-6H2,1-3H3,(H,17,18)

Inchi Key

FZQHRSMRHOQSOY-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library