Skip to Content
MilliporeSigma

BDPH9BCDCB26

Bide Pharmatech

tert-Butyl (tetrahydro-2H-thiopyran-4-yl)carbamate

Synonym(s):

tert-butyl tetrahydro-2H-thiopyran-4-ylcarbamate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H19O2N1S1
Molecular Weight:
217.33
MDL number:


Inchi Code

1S/C10H19NO2S/c1-10(2,3)13-9(12)11-8-4-6-14-7-5-8/h8H,4-7H2,1-3H3,(H,11,12)

Inchi Key

IRZQBMIZGJSXFN-UHFFFAOYSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library