Skip to Content
MilliporeSigma

BDPH9BCDD03B

Bide Pharmatech

(2-Fluoro-4,6-dimethoxyphenyl)boronic acid

Synonym(s):

(2-fluoro-4,6-dimethoxyphenyl)boronic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H10O4F1B1
CAS Number:
Molecular Weight:
199.97
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H10BFO4/c1-13-5-3-6(10)8(9(11)12)7(4-5)14-2/h3-4,11-12H,1-2H3

Inchi Key

VRYIYLGRLZLYPA-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library