Skip to Content

BDPH9BCDD596

Bide Pharmatech

3-Bromo-6-methoxy-2-methylimidazo[1,2-b]pyridazine

Synonym(s):

3-bromo-6-methoxy-2-methylimidazo[1,2-b]pyridazine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H8O1N3Br1
CAS Number:
Molecular Weight:
242.08
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H8BrN3O/c1-5-8(9)12-6(10-5)3-4-7(11-12)13-2/h3-4H,1-2H3

Inchi Key

FHRDIUPGIHSDDW-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Yellow solid



Still not finding the right product?


signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library