Skip to Content

BDPH9BCDDEA0

Bide Pharmatech

tert-Butyl 2-(2-(prop-2-yn-1-yloxy)ethoxy)acetate

Synonym(s):

tert-butyl 2-(2-(prop-2-yn-1-yloxy)ethoxy)acetate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C11H18O4
CAS Number:
Molecular Weight:
214.26
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C11H18O4/c1-5-6-13-7-8-14-9-10(12)15-11(2,3)4/h1H,6-9H2,2-4H3

Inchi Key

UKKQWNXHEGQLMK-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Liquid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library