Inchi Code
1S/C10H17N/c1-2-3-8-11-9-6-4-5-7-10-11/h1H,3-10H2
Inchi Key
OQBUHCAAQPHPCG-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
95%
Storage temp.
Inert atmosphere, 2-8C
Country of Origin
CN
Physical Form
Liquid
Still not finding the right product?
Explore all of our products under 1-(But-3-yn-1-yl)azepane
hstatements
Signal Word
Danger
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

