Skip to Content
MilliporeSigma

BDPH9C01F0C7

Bide Pharmatech

(S)-3-Amino-2-methylbutan-2-ol hydrochloride

Synonym(s):

(3S)-3-amino-2-methyl-2-butanol hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C5H14O1N1Cl1
CAS Number:
Molecular Weight:
139.62
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C5H13NO.ClH/c1-4(6)5(2,3)7;/h4,7H,6H2,1-3H3;1H/t4-;/m0./s1

Inchi Key

YURXCUVDCIDDHM-WCCKRBBISA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library