Skip to Content
MilliporeSigma

BDPH9C01F6A0

Bide Pharmatech

(R)-1-(3-(Difluoromethyl)-2-fluorophenyl)ethan-1-amine hydrochloride

Synonym(s):

(R)-1-(3-(difluoromethyl)-2-fluorophenyl)ethan-1-amine hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H11N1Cl1F3
CAS Number:
Molecular Weight:
225.64
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H10F3N.ClH/c1-5(13)6-3-2-4-7(8(6)10)9(11)12;/h2-5,9H,13H2,1H3;1H/t5-;/m1./s1

Inchi Key

LMVWLMYIWIMIAX-NUBCRITNSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?


Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library