Skip to Content
MilliporeSigma

BDPH9C01F7C3

Bide Pharmatech

2-Cyclopropylbenzaldehyde

Synonym(s):

2-cyclopropylbenzaldehyde

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H10O1
CAS Number:
Molecular Weight:
146.19
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C10H10O/c11-7-9-3-1-2-4-10(9)8-5-6-8/h1-4,7-8H,5-6H2

Inchi Key

ZVDXHSUCDWVQEY-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Liquid



Still not finding the right product?

Explore all of our products under 2-Cyclopropylbenzaldehyde



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library