Skip to Content
MilliporeSigma

BDPH9C0201B2

Bide Pharmatech

(S)-4-Phenyl-2-(pyrazin-2-yl)-4,5-dihydrooxazole

Synonym(s):

(S)-4-phenyl-2-(pyrazin-2-yl)-4,5-dihydrooxazole

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C13H11O1N3
CAS Number:
Molecular Weight:
225.25
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C13H11N3O/c1-2-4-10(5-3-1)12-9-17-13(16-12)11-8-14-6-7-15-11/h1-8,12H,9H2/t12-/m1/s1

Inchi Key

LSZGSJFTOXBLGG-GFCCVEGCSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Keep in dark place, inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Colorless to Yellow Liquid or Semi-Solid or solid



Still not finding the right product?


Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library