Skip to Content
MilliporeSigma

BDPH9C021C10

Bide Pharmatech

(S)-3-((tert-Butoxycarbonyl)amino)-4-methylpentanoic acid

Synonym(s):

(3S)-3-[(tert-butoxycarbonyl)amino]-4-methylpentanoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C11H21O4N1
CAS Number:
Molecular Weight:
231.29
MDL number:


Inchi Code

1S/C11H21NO4/c1-7(2)8(6-9(13)14)12-10(15)16-11(3,4)5/h7-8H,6H2,1-5H3,(H,12,15)(H,13,14)/t8-/m0/s1

Inchi Key

LUXMZCJCTUATDM-QMMMGPOBSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Form: powder Colour: white to pale-yellow



Still not finding the right product?


signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library