Inchi Code
1S/C9H9F4N.ClH/c1-5(14)6-2-3-7(8(10)4-6)9(11,12)13;/h2-5H,14H2,1H3;1H/t5-;/m1./s1
Inchi Key
NLBPCMJOACOYKH-NUBCRITNSA-N
Shipping Temperature
Normal
Purity
95%
Storage temp.
Inert atmosphere, room temperature
Country of Origin
CN
Physical Form
Solid
Still not finding the right product?
Explore all of our products under (R)-1-(3-Fluoro-4-(trifluoromethyl)phenyl)ethanamine hydrochloride
hstatements
signalword
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
