Skip to Content
MilliporeSigma

BDPH9C02254E

Bide Pharmatech

(R)-3-Amino-1-methylpiperidin-2-one hydrochloride

Synonym(s):

(R)-3-amino-1-methylpiperidin-2-one hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H13O1N2Cl1
CAS Number:
Molecular Weight:
164.63
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C6H12N2O.ClH/c1-8-4-2-3-5(7)6(8)9;/h5H,2-4,7H2,1H3;1H/t5-;/m1./s1

Inchi Key

HHUWUTGRVZJMSM-NUBCRITNSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library