Skip to Content
MilliporeSigma

BDPH9C022AA5

Bide Pharmatech

MEthyl (2s)-2-amino-3-(furan-2-yl)propanoate

Synonym(s):

methyl (2S)-2-amino-3-(2-furyl)propanoate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H11O3N1
CAS Number:
Molecular Weight:
169.18
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H11NO3/c1-11-8(10)7(9)5-6-3-2-4-12-6/h2-4,7H,5,9H2,1H3/t7-/m0/s1

Inchi Key

RGZJZYPBZXARCU-ZETCQYMHSA-N

Shipping Temperature

Cold-chain transportation

Purity

95%

Storage temp.

Keep in dark place, sealed in dry, store in freezer, under -20C

Country of Origin

CN

Physical Form

Sticky Liquid to Solid



Still not finding the right product?


hstatements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library