Skip to Content
MilliporeSigma

BL3H1F1C8A1D

BLD Pharmatech

Pyridin-3-ol

Synonym(s):

3(2H)-pyridinone

Select a Size

Change View

About This Item

Linear Formula:
C5H5O1N1
CAS Number:
Molecular Weight:
95.1
MDL number:
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C5H5NO/c7-5-2-1-3-6-4-5/h1-4,7H

Inchi Key

GRFNBEZIAWKNCO-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Keep in dark place, sealed in dry, room temperature

Country of Origin

CN

Physical Form

Form: Crystal - Powder / Colour: White - Yellow red - Light-red

Melting Point

125-128C



Still not finding the right product?

Explore all of our products under Pyridin-3-ol


hstatements

Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library