Skip to Content
MilliporeSigma

Skip To

BL3H97A47BB2

BLD Pharmatech

Ethyl 3-bromo-4-fluorobenzoate

Synonym(s):

ethyl 3-bromo-4-fluorobenzoate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 g
Ships in 5 days - from Aldrich Partner.
$10.00
5 g
Ships in 5 days - from Aldrich Partner.
$14.00
10 g
Ships in 5 days - from Aldrich Partner.
$25.00
25 g
Ships in 5 days - from Aldrich Partner.
$40.00
100 g
Ships in 5 days - from Aldrich Partner.
$121.00

About This Item

Linear Formula:
C9H8O2Br1F1
CAS Number:
Molecular Weight:
247.06
MDL number:

Vendor SKU: BD240149-1G

$10.00


Ships in 5 days - from Aldrich Partner.

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H8BrFO2/c1-2-13-9(12)6-3-4-8(11)7(10)5-6/h3-5H,2H2,1H3

Inchi Key

FDVAAQDTTZNHKG-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

White to yellow powder or crystals



Still not finding the right product?

Explore all of our products under Ethyl 3-bromo-4-fluorobenzoate


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library