Skip to Content
MilliporeSigma

NMIM924

(-)-Ephedrine hydrochloride

NMI Australia

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C10H16NOCl
CAS Number:
Molecular Weight:
201.69
UNSPSC Code:
41116107
EC Number:
200-074-6
eCl@ss:
25140904
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


agency

NMI Australia certified reference material

manufacturer/tradename

NMI Australia

format

neat

storage temp.

room temp

InChI

1S/C10H15NO.ClH/c1-8(11-2)10(12)9-6-4-3-5-7-9;/h3-8,10-12H,1-2H3;1H/t8-,10-;/m0./s1

InChI key

BALXUFOVQVENIU-GNAZCLTHSA-N

General description

Ephedrine is a methamphetamine analog commonly used as a stimulant and appetite suppressant. It is also found as an active constituent in herbs used for traditional Chinese medicine, such as má huáng. This reference material can be used as a starting material in the preparation of calibrators or controls for clinical or forensic applications or in quality control testing of dietary supplements containing ephedrine.


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Acute Tox. 4 Oral - STOT SE 3

Target Organs

Central nervous system

Storage Class

11 - Combustible Solids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library