Skip to Content
MilliporeSigma

NMIP1801

α-Zearalanol

NMI Australia

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C18H26O5
CAS Number:
Molecular Weight:
322.40
UNSPSC Code:
41116107
EC Number:
247-769-0
eCl@ss:
25140904
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

agency

NMI Australia certified reference material

form

solid

manufacturer/tradename

NMI Australia

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

neat

storage temp.

2-8°C

InChI

1S/C18H26O5/c1-12-6-5-9-14(19)8-4-2-3-7-13-10-15(20)11-16(21)17(13)18(22)23-12/h10-12,14,19-21H,2-9H2,1H3/t12-,14?/m0/s1

InChI key

DWTTZBARDOXEAM-NBFOIZRFSA-N



Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Codes

Hazard Classifications

Repr. 2 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library