Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C25H40O8
CAS Number:
Molecular Weight:
468.58
UNSPSC Code:
41116107
eCl@ss:
25140904
agency
NMI Australia certified reference material
manufacturer/tradename
NMI Australia
format
neat
storage temp.
2-8°C
Storage Class
11 - Combustible Solids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.