Skip to Content
MilliporeSigma

COM638465752

Combi-Blocks, Inc.

7-Chloro-6-methyl-1H-benzimidazole

Synonym(s):

7-chloro-6-methyl-1H-benzimidazole

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H7N2Cl1
CAS Number:
Molecular Weight:
166.61
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H7ClN2/c1-5-2-3-6-8(7(5)9)11-4-10-6/h2-4H,1H3,(H,10,11)

Inchi Key

JRTXYNGGGOWIBM-UHFFFAOYSA-N

Purity

98%

Country of Origin

CN





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library