Skip to Content
MilliporeSigma

COMH93D67E2A

Combi-Blocks, Inc.

Ospemifene

Synonym(s):

2-{4-[(1Z)-4-chloro-1,2-diphenyl-1-butenyl]phenoxy}ethanol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C24H23O2Cl1
CAS Number:
Molecular Weight:
378.9
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C24H23ClO2/c25-16-15-23(19-7-3-1-4-8-19)24(20-9-5-2-6-10-20)21-11-13-22(14-12-21)27-18-17-26/h1-14,26H,15-18H2/b24-23-

Inchi Key

LUMKNAVTFCDUIE-VHXPQNKSSA-N

Purity

96%

Country of Origin

CN



Still not finding the right product?

Explore all of our products under Ospemifene


hstatements



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library