Skip to Content
MilliporeSigma

348

Rose Bengal

Harleco®, suitable for detection

Synonym(s):

Rose Bengal, Aniline Rose

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H2Cl4I4Na2O5
CAS Number:
Molecular Weight:
1017.64
Colour Index Number:
45440
UNSPSC Code:
41116121
EC Index Number:
211-183-3
MDL number:
eCl@ss:
25140910
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


manufacturer/tradename

Harleco®

Quality Segment

application(s)

detection

shipped in

ambient

InChI

1S/C20H4Cl4I4O5.2Na/c21-9-7-8(10(22)12(24)11(9)23)20(33-19(7)31)3-1-5(25)15(29)13(27)17(3)32-18-4(20)2-6(26)16(30)14(18)28;;/h1-2,29-30H;;/q;2*+1/p-2

InChI key

KCQREHTWEUECQT-UHFFFAOYSA-L

General description

HARLECO® Rose Bengal
348-12 25g Glass Bottle
HARLECO® Used to detect bacteria in soil samples

Application

Certified by the Biological Stain Commission (BSC)
For In-Vitro Diagnostic Use.
For Professional Use.
Used for bacteria in soil.
HARLECO® Used to detect bacteria in soil samples

Legal Information

HARLECO is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Rose Bengal


Storage Class

10-13 - German Storage Class 10 to 13



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library