Skip to Content
MilliporeSigma

9Q0416

Anti-Barbituate (10mg) KC

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Clone:
monoclonal
Technique(s):
ELISA: suitable
Application:
ELISA
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


antibody form

purified antibody

Quality Segment

clone

monoclonal

species reactivity

mouse, all

concentration

(Please refer to lot specific datasheet.)

technique(s)

ELISA: suitable

isotype

IgG1

General description

Barbiturates are a class of controlled drug that act as suppressants for the central nervous system. Both phycologically and physically addictive barbiturates are often abused for recreational use and are frequently included in basic drug screens

Application

Suggested antibody pairs MABX4081-1KC and MABX4082-1KC

Preparation Note

2-8°C. For long-term storage, freeze at -20°C.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Explore all of our products under Anti-Barbituate (10mg) KC


Storage Class

12 - Non Combustible Liquids

WGK

WGK 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library