Skip to Content
MilliporeSigma

FR139475082

K-080 Green Std PS 10%

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

polystyrene

Quality Segment

diam.

0.750—0.890 μm

Application

Dyed microspheres are extremely uniform and are dyed internally (their surface properties are unchanged), insuring maximum color brilliance and preventing dye leaching in water or aqueous buffers.


Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library