Skip to Content
MilliporeSigma

PG9H9BB386AD

PrecisePEG

DOTA-NHS Ester

Synonym(s):

2,2',2''-(10-(2-((2,5-dioxopyrrolidin-1-yl)oxy)-2-oxoethyl)-1,4,7,10-tetraazacyclododecane-1,4,7-triyl)triacetic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C20H31O10N5
CAS Number:
Molecular Weight:
501.49
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C20H31N5O10/c26-15-1-2-16(27)25(15)35-20(34)14-24-9-7-22(12-18(30)31)5-3-21(11-17(28)29)4-6-23(8-10-24)13-19(32)33/h1-14H2,(H,28,29)(H,30,31)(H,32,33)

Inchi Key

XSVWFLQICKPQAA-UHFFFAOYSA-N

Shipping Temperature

Cold Pack

Purity

90%

Storage temp.

-20

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under DOTA-NHS Ester


hstatements

Precautionary Statements



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library