Skip to Content
MilliporeSigma

PG9H9BC7B112

PrecisePEG

Aminooxy-PEG3-Azide

Synonym(s):

O-(2-(2-(2-(2-azidoethoxy)ethoxy)ethoxy)ethyl)hydroxylamine

Select a Size

Change View

Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H18N4O4/c9-12-11-1-2-13-3-4-14-5-6-15-7-8-16-10/h1-8,10H2

Inchi Key

WSRXFPCTWIFEOF-UHFFFAOYSA-N

Storage temp.

-20

Purity

96%

Country of Origin

CN

Shipping Temperature

ambient

Physical Form

liquid



Still not finding the right product?

Explore all of our products under Aminooxy-PEG3-Azide


MSDS

https://precisepeg.com/cdn/shop/files/AG-1723_SDS.pdf

hstatements

Precautionary Statements



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library