Skip to Content
MilliporeSigma

PG9H9BC7B166

PrecisePEG

endo BCN-PEG3-NHS Ester

Synonym(s):

2,5-dioxopyrrolidin-1-yl 1-((1R,8S,9s)-bicyclo[6.1.0]non-4-yn-9-yl)-3-hydroxy-2,7,10,13-tetraoxa-4-azahexadec-3-en-16-oate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View



Inchi Code

1S/C24H34N2O9/c27-21-7-8-22(28)26(21)35-23(29)9-11-31-13-15-33-16-14-32-12-10-25-24(30)34-17-20-18-5-3-1-2-4-6-19(18)20/h18-20H,3-17H2,(H,25,30)/t18-,19+,20-

Inchi Key

HCELGENRAWFQSJ-ACDBMABISA-N

Storage temp.

-20

Purity

90%

Country of Origin

CN

Shipping Temperature

ambient

Physical Form

Yellow



MSDS

https://precisepeg.com/cdn/shop/files/AG-2012_SDS.pdf

hstatements

pcodes



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library