Skip to Content
MilliporeSigma

08088

p-Aminohippuric acid

puriss., ≥99.0% (T)

Synonym(s):

4-Aminohippuric acid, N-(4-Aminobenzoyl)glycine

Select a Size

Change View

About This Item

Linear Formula:
NH2C6H4C(O)NHCH2COOH
CAS Number:
Molecular Weight:
194.19
EC Number:
200-518-9
UNSPSC Code:
12352200
MDL number:
Beilstein/REAXYS Number:
1213676
eCl@ss:
32150312
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

puriss.

assay

≥99.0% (T)

mp

199-200 °C (dec.) (lit.), 200-202 °C

SMILES string

Nc1ccc(cc1)C(=O)NCC(O)=O

InChI

1S/C9H10N2O3/c10-7-3-1-6(2-4-7)9(14)11-5-8(12)13/h1-4H,5,10H2,(H,11,14)(H,12,13)

InChI key

HSMNQINEKMPTIC-UHFFFAOYSA-N



Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library