Skip to Content
MilliporeSigma

PMC2011

BippyPhos

≥98%, ISOM8™

Synonym(s):

5-(Di-tert-butylphosphino)-1′, 3′, 5′-triphenyl-1′H-[1,4′]bipyrazole, 5-[bis(1,1-dimethylethyl)phosphino]-1′,3′,5′-triphenyl-1,4′-Bi-1H-pyrazole, t-Bu-BippyPhos, BippyPhos

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C32H35N4P
CAS Number:
Molecular Weight:
506.62
UNSPSC Code:
12352005
MDL number:
eCl@ss:
32150313
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

ISOM8™

Quality Segment

assay

≥98%

form

solid

purified by

HPLC

reaction suitability

reagent type: ligand, reagent type: ligand
reaction type: Amidations, reagent type: ligand
reaction type: Buchwald-Hartwig Cross Coupling Reaction, reagent type: ligand
reaction type: Cross Couplings

mp

169-173 °C

application(s)

diagnostic assay manufacturing
life science and biopharma

functional group

phosphine

storage temp.

room temp

SMILES string

CC(C)(C)P(c1ccnn1-c2c(nn(-c3ccccc3)c2-c4ccccc4)-c5ccccc5)C(C)(C)C

InChI

1S/C32H35N4P/c1-31(2,3)37(32(4,5)6)27-22-23-33-36(27)30-28(24-16-10-7-11-17-24)34-35(26-20-14-9-15-21-26)29(30)25-18-12-8-13-19-25/h7-23H,1-6H3

InChI key

PTXJGGGNGMPMBG-UHFFFAOYSA-N

General description

Welcome to our ISOM8™ line of high-quality products available and able to scale for manufacturing. Within our ISOM8 team, we have expertise and documentation to support ISO 9001 industrial and non-GMP pharma production needs, as defined by our M-Clarity™ Program.Our portfolio of ISOM8 products and chemistries is ever evolving to support our valued Sigma-Aldrich® customers′ requirements. Please navigate to our documents section to look at quality documentation for this product, provided with support of our Elevate Program.

Application

The BippyPhos is a highly versatile, bulky, and electron-rich bipyrazole phosphine ligand. It excels in palladium-catalyzed cross-coupling reactions — most notably Buchwald-Hartwig amination — and is prized for its exceptional compatibility with challenging substrates, including electron-deficient aryl chlorides and ammonia.


Notable Characteristics & Key Uses


  • Chemical Structure: BippyPhos is a bipyrazole ligand featuring a di-tert-butylphosphine moiety that imparts high electron density, significant steric bulk, and strong sigma-donor character to the palladium center.
  • Broad Solubility: BippyPhos exhibits excellent solubility in a wide range of common organic solvents, enabling its deployment across diverse reaction platforms and scales.
  • Steric and Electronic Synergy: The pronounced steric shielding of BippyPhos accelerates the critical reductive elimination step in catalytic cycles, directly improving turnover frequency and product selectivity.
  • Air Stability: BippyPhos is air-stable and straightforward to handle, making it well-suited for large-scale industrial applications.
  • Versatility: Beyond C-N coupling, BippyPhos is equally effective for C-O and C-C bond-forming reactions, demonstrating broad applicability across coupling chemistries.
  • Buchwald-Hartwig Amination (C-N Coupling): Particularly effective for coupling primary amines with aryl halides, including electron-deficient aryl chlorides. Widely used in the preparation of pharmaceutical intermediates and demonstrates exceptional efficiency for the monoarylation of ammonia and hydrazine.
  • Suzuki-Miyaura and Heck Coupling: BippyPhos stabilizes active palladium complexes, markedly improving efficiency and selectivity in C-C bond-forming reactions.
  • C-O Coupling: Functions as an efficient catalyst for coupling primary alcohols with (hetero)aryl halides.
  • Hydroxylation: Employed in palladium-catalyzed hydroxylation of aryl halides to afford synthetically valuable phenols.
  • Coordination Chemistry: Utilized in the investigation of metal-ligand interactions and the development of novel functional materials.

Recent Studies and Innovations 


  1. Mechanochemical Synthesis: Mechanochemical methodologies have been applied to generate active Pd/BippyPhos catalysts in a solvent-free medium, successfully coupling aryl halides with amines in high yield. This approach underscores BippyPhos′s versatility beyond conventional solution-phase chemistry (Deniz et al., 2025).
  2. Amination under Mild Conditions: Pd-BippyPhos complexes demonstrate remarkable efficacy in the amination of aryl fluorosulfates, affording biarylamines in excellent yields under mild conditions - surpassing alternative ligand systems tested under identical reaction parameters (Karthik S et al., 2024).

Legal Information

ISOM8 is a trademark of Sigma-Aldrich Co. LLC
M-CLARITY is a trademark of Merck KGaA, Darmstadt, Germany
SIGMA-ALDRICH is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library