Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Linear Formula:
N=N=CHCOOC2H5
CAS Number:
Molecular Weight:
114.10
EC Number:
210-810-8
UNSPSC Code:
12352100
PubChem Substance ID:
Beilstein/REAXYS Number:
107654
MDL number:
grade
purum
impurities
≤10% dichloromethane
refractive index
n
bp
140-141 °C/720 mmHg (lit.)
mp
−22 °C (lit.)
density
1.085 g/mL at 25 °C (lit.)
storage temp.
2-8°C
SMILES string
CCOC(=O)C=[N+]=[N-]
InChI
1S/C4H6N2O2/c1-2-8-4(7)3-6-5/h3H,2H2,1H3
InChI key
YVPJCJLMRRTDMQ-UHFFFAOYSA-N
Application
Other Notes
Reagent for the cyclopropanation of olefins
Signal Word
Warning
Hazard Codes
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3
Target Organs
Central nervous system
Storage Class
4.1B - Flammable solid hazardous materials
WGK
WGK 3
Flash Point (°F)
116.6 °F - closed cup
Flash Point (°C)
47 °C - closed cup
PPE (personal protective equipment)
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


