Skip to Content
MilliporeSigma

APREST89757

PrEST Antigen UTP6

Prestige Antigens™ Powered by Atlas Antibodies, buffered aqueous solution

Synonym(s):

C17orf40, HCA66

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160702
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


recombinant

expressed in E. coli

assay

>80% (SDS-PAGE)

form

buffered aqueous solution

mol wt

predicted mol wt 33 kDa

purified by

immobilized metal affinity chromatography (IMAC)

concentration

≥0.5 mg/mL

immunogen sequence

AKSQEDTEAVFKKALLAVIGADSVTLKNKYLDWAYRSGGYKKARAVFKSLQESRPFSVDFFRKMIQFEKEQESCNMANIREYYERALREFGSADSDLWMDYMKEELNHPLGRPENCGQIYWRAMKMLQG

Ensembl | human accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... UTP6(55813)

General description

Recombinant protein fragment of Human UTP6 with N-terminal His6ABP (ABP = Albumin Binding Protein derived from Streptococcal Protein G) fusion tag.    

Application

Blocking agent and positive assay control using corresponding antibodies.

Physical form

Solution in phosphate-buffered saline and 1M Urea, pH 7.4

Preparation Note

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user.

Other Notes

Corresponding Antibody HPA055806.

Legal Information

Prestige Antigens is a trademark of Sigma-Aldrich Co. LLC


Storage Class

10 - Combustible liquids

WGK

WGK 2

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library