MilliporeSigma
BCS-262CS
CoolCell™ SV10
Stem Cell Cryopreservation System
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
41103000
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
feature
for vials 10 mL, Includes XT Starter cooling base with CoolRack SV10 and CoolCell SV10 cell freezing container
manufacturer/tradename
BioCision BCS-262CS
General description
CoolCell SV10 Stem Cell Cryopreservation System is a kit designed for efficient and controlled preparation and cryopreservation of stem cells and cell therapy products.
Ice-free cell preparation:
The XT Starter ice-free cooling base and CoolRack SV10 vial module provide up to 4 hours of ice-free cooling (0.5 to 4.0°C) for cell and cryoprotectant preparation. Simply freeze the XT Starter core in a -20°C freezer then rest the thermo-conductive CoolRack SV10 on top. The system will equilibrate to approximately 2°C and keep all samples uniformly cold and free of ice contamination. The small footprint and low profile make this an ideal cooling solution for cell preparation performed in a tissue culture a hood.
Dimensions:
XT Starter ice-free cooling base: (L x W x H): 16.2 x 12.0 x 5.5 cm /CoolRack SV10 module: (L x W x H): 12.7 x 8.6 x 3.8 cm; Well dimensions: (Diam x height): 23.6 x 48 mm; Holds 12 vials.
Controlled-rate cell freezing:
The CoolCell SV10 cell freezing container provides highly reproducible -1°C/minute freezing rate for cells when cryopreserved in a -80°C freezer. No programming or alcohol is required - simply place in a -80°C freezer. The insulative outer housing and thermo-conductive core ensure consistent heat removal to all vials throughout the freezing period. CoolCell containers are unbreakable, open easily when frozen, and are not cold to the touch when frozen - no "frosty" fingers.
Holds 6 injectable vials.
Legal Information
CoolCell is a trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)