Skip to Content
MilliporeSigma

BPS78856

U2 Assay Buffer

Synonym(s):

buffers

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
47101613
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


manufacturer/tradename

BPS Bioscience

storage temp.

-65 to -96°C

General description

U2 Assay Buffer is an assay buffer optimized for use with the BPS Bioscience Intrachain TR-FRET Assay Kits and Ubi-Mix (BPS Bioscience #82185).

Application

Screen molecules that inhibit ubiquitin ligase activity in drug discovery high throughput screening (HTS) applications, determine compound IC50 and perform real-time kinetics analyses in Intrachain TR-FRET assays.

Datasheet and Protocols may be found here.

Preparation Note

This is a ready to use assay buffer.Aqueous solution.

Analysis Note

Avoid freeze/thaw cycles.


Storage Class

10 - Combustible liquids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library