Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.81
UNSPSC Code:
41106514
eCl@ss:
32160703
Biological source:
human foreskin (juvenile epidermis)
Morphology:
(epidermal)
biological source
human foreskin (juvenile epidermis)
packaging
pkg of 500,000 cells
morphology
(epidermal)
technique(s)
cell culture | mammalian: suitable
shipped in
ambient
storage temp.
room temp
General description
Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.
Application
Primary Normal Human Epidermal Melanocytes (NHEM) are isolated from the epidermis of juvenile foreskin and are provided in a proliferating format. Melanocytes are located in the stratum basale, but branch out between the keratinocytes in suprabasal layers. About 5-10 % of the cells in the epidermis are melanocytes. The main purpose of melanocytes is the production of melanin, the protein responsible for the pigmentation of the skin, eyes, and hair. Melanin protects the cells in the skin and in deeper layers from the hazardous effects of UV radiation. It is produced and stored in melanosomes, which are located close to the melanocyte membrane, but it can also be transferred to neighboring keratinocytes.Normal Human Epidermal Melanocytes are isolated using the serum-free, PMA (Phorbol Myristate Acetate)-containing Melanocyte Growth Medium. Since PMA is a tumor promoting mitogen, it can interfere with experimental approaches. Therefore, we recommend using cells isolated in Melanocyte Growth Medium M2 and also using this medium for cultivation. Normal Human Epidermal Keratinocytes (NHEK) and Normal Human Dermal Fibroblasts (NHDF) from the same donor are available on request.
Biochem/physiol Actions
Skin
Preparation Note
Click here for more information.
Analysis Note
Rigid quality control tests are performed for each lot of Normal Human Epidermal Melanocytes. They are tested for cell morphology, adherence rate and cell viability. Furthermore, immunohistochemical tests for the cell-type specific marker, Mel-5, are carried out for each lot. Growth performance is tested through multiple passages up to 15 population doublings (PD) under culture conditions without antibiotics and antimycotics. In addition, all cells have been tested for the absence of HIV-1, HIV-2, HBV, HCV, HTLV-1, HTLV-2 and microbial contaminants (fungi, bacteria, and mycoplasma).
Other Notes
PMA-containing Melanocyte Growth Medium
Recommended Plating Density: 5000 - 10000 cells per cm2Passage After Thawing: P2Tested Markers: Mel-5 positiveGuaranteed population doublings: >15
Disclaimer
Although tested negative for HIV-1, HIV-2, HBV, HCV, HTLV-1 and HTLV-2, the cells – like all products of human origin – should be handled as potentially infectious. No test procedure can completely guarantee the absence of infectious agents.
This product is regulated in France when intended for scientific purposes, including import, export, and conservation activities (Articles L.1211-1, paragraph 2, and L.1243-4 of the French Public Health Code). The purchaser (i.e. end user) is required to obtain an import authorization from the France Ministry responsible for Research (as referenced in Article L.1245-5-1 II of the French Public Health Code) or a conservation authorization from the same ministry (as referenced in Article L.1243-4 of the French Public Health Code). By placing an order for this product, you confirm that you have obtained the appropriate import or conservation authorization for the product. For more information regarding the use of human biological samples for research purposes in France, please contact the relevant French authorities.
Still not finding the right product?
Explore all of our products under Normal Human Epidermal Melanocytes (NHEM)
Storage Class
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point (°F)
No data available
Flash Point (°C)
No data available
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.