Skip to Content
MilliporeSigma

IX-2H7-E10 Mouse x Mouse Hybridoma, ECACC 08041722

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41106514
eCl@ss:
32160701
Biological source:
Mouse x Mouse Hybridoma
Isotype:
IgG1
Technique(s):
cell culture | mammalian: suitable
Myeloma:
NS-1 (P3-X63/Ag8-NS1)
Reactivity:
Anti-L. (L) amazonensis-less with L. major
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

IX-2H7-E10, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

biological source

Mouse x Mouse Hybridoma

manufacturer/tradename

ECACC

packaging

tube of 5 μg 08041722-DNA-5UG, pkg of vial of cells 08041722-1VL

ImmunoDon

BALB/c mouse spleen

Myeloma

NS-1 (P3-X63/Ag8-NS1)

Reactivity

Anti-L. (L) amazonensis-less with L. major

technique(s)

cell culture | mammalian: suitable

isotype

IgG1

shipped in

dry ice

storage temp.

−196°C

Immunogen

Membrane from Leishmania (L) amazonensis WR303 strain MHOM/BR/68/H6

Application

Detection of L. amazonensis

Biochem/physiol Actions

IX-2H7-E10 was developed by immunizing mice with membranes from L. mexicana amazonensis and fusing the mouse spleen cells with NS-1(P3-X63/Ag8-NS1)
Mouse NS1 x BALB/c mouse

Preparation Note

RPMI 1640 + 2mM glutamine + 10% Foetal Bovine Serum (FBS)
Seed cultures between 1-3x100,000 cells/ml; Saturation density 1,000,000 5% CO2; 37°C

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
This cell line is part of the European Collection of Authenticated Cell Cultures (ECACC), an international repository managed by the United Kingdom Health Security Agency (UKHSA). No licensing agreement is required when either this cell line or the DNA extracted from it are used for internal research purposes only. Any other use of these products is prohibited without the express written permission of UKHSA. Inquiries regarding authorized use of this cell line or its genomic DNA may be directed to culturecollections@ukhsa.gov.uk.




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library