Skip to Content
MilliporeSigma

CLSETAT04

Corning® Gosselin™ T Shaped Spreader

Green PS, sterile

Synonym(s):

Agar Spreader, Cell Spreader T-Type, T Rod Spreader

Select a Size

Change View

About This Item

UNSPSC Code:
41122100
eCl@ss:
32039016
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

polystyrene

sterility

sterile

packaging

case of 3500

manufacturer/tradename

Corning ETAT-04

General description

Disposable pipettes designed for accurate liquid transfer in laboratory applications, offering reliable performance for sampling, dosing, and general lab use.

Application

Microbiology / Plating

Legal Information

Corning is a registered trademark of Corning, Inc.
Gosselin is a trademark of Corning, Inc.




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library