Skip to Content
MilliporeSigma

CLSTAT30C01

Corning® Gosselin™ Dipper

capacity 40 mL, with Removable Handle, Blue PP with Blue Screw Cap

Synonym(s):

Collection Dipper, Liquid Scoop, Sampling Dipper

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41104118
eCl@ss:
27270911
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

polypropylene

sterility

non-sterile

feature

cap, Blue PP with Blue Screw Cap, with Removable Handle

packaging

case of 250

manufacturer/tradename

Corning TAT30C-01

capacity

40 mL

General description

Sampling dippers with removable handles for liquid collection, designed for hygienic sampling in environmental, food, and laboratory applications.

Application

Sample Collection

Legal Information

Corning is a registered trademark of Corning, Inc.
Gosselin is a trademark of Corning, Inc.


Still not finding the right product?

Explore all of our products under Corning® Gosselin Dipper




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library