Skip to Content
MilliporeSigma

F0193

Fetal Bovine Serum

Uruguayan or Brazilian origin, sterile, suitable for cell culture

Synonym(s):

FBS, FCS, sera, serum

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352200
MDL number:
eCl@ss:
32160701
Biological source:
bovine fetus
Sterility:
sterile
Form:
liquid
Shipped in:
dry ice
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bovine fetus

Quality Segment

sterility

sterile

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

haemoglobin < 30 mg/100mL, <30 mL endotoxin

pH

6-8

shipped in

dry ice

storage temp.

−20°C

Application

FBS is used in a broad range of cell culture applications.

Analysis Note

  • Endotoxin and hemoglobin tested
  • Endotoxin and hemoglobin tested
  • Tested for the presence of aerobic and anaerobic bacteria, fungi, yeast and Mycoplasma
  • Triple filtered with 0.1 µm membrane under aseptic conditions

Legal Information

PRODUCT AVAILABLE ONLY FOR CHINA


Still not finding the right product?

Explore all of our products under Fetal Bovine Serum


Storage Class

10 - Combustible liquids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library