Skip to Content
MilliporeSigma

OGS1147

PSF-CMV-PURO-NH2-GST-3C - N-TERMINAL GST TAG MAMMALIAN PLASMID

plasmid vector for molecular cloning

Synonym(s):

cloning vector, expression vector, molecular cloning vector, plasmid, plasmid vector, snapfast vector, vector

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.85
UNSPSC Code:
12352200
Promoter:
Promoter name: CMV
Promoter activity: constitutive
Promoter type: mammalian
Origin of replication:
pUC (500 copies)
Bacteria selection:
kanamycin
Reporter gene:
none
Peptide cleavage:
3C
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


tag

GST tagged

form

buffered aqueous solution

mol wt

size 6827 bp

bacteria selection

kanamycin

mammalian cells selection

puromycin

origin of replication

pUC (500 copies)

peptide cleavage

3C

peptide tag location

N-terminal

promoter

Promoter name: CMV
Promoter activity: constitutive
Promoter type: mammalian

reporter gene

none

shipped in

ambient

storage temp.

−20°C

General description

This plasmid is designed to express tagged proteins in mammalian cells either by transient transfection or by creating stable cell lines. It contains a puromycin resistance expression cassette using the human Ubiquitin promoter to drive expression and allow for the selection of cells containing the plasmid.

About the Peptide Tag:This plasmid contains an n-terminal Glutathione-S-Transferase (GST) affinity tag that can be fused to a gene of interest to allow protein detection and/or purification. The sequence of the tag is: SPILGYWKIKGLVQPTRLLLEYLEEKYEEHLYERDEGDKWRNKKFELGLEFPNLPYYIDGDVKLTQSMAIIRYIADKHNMLGGCPKERAEISMLEGAVLDIRYGVSRIAYSKDFETLKVDFLSKLPEMLKMFEDRLCHKTYLNGDHVTHPDFMLYDALDVVLYMDPMCLDAFPKLVCFKKRIEAIPQIDKYLKSSKYIAWPLQGWQATFGGGDHPPKS.

About the Cleavage Tag:This plasmid also encodes a protease cleavage site that is designed to be positioned between your gene of interest and the tag to allow the removal of the tag following protein purification or isolation. This plasmid contains a 3C cleavage tag. The protein sequence of the cleavage tag is: LEVLFQGP. Human Rhinovirus (HRV) 3C Protease is a highly specific protease that cleaves between the Glu and Gly residues of its recognition site. It is often produced with the trademname ′PreScission protease′.

Promoter Expression Level: This plasmid contains the mammalian CMV promoter to drive gene expression. We have tested all of our mammalian promoters in a range of cell types and CMV is consistently the strongest in those we have studied. However there are many reports of the CMV promoter demonstrating silencing by methylation in long-term culture.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.oxgene.com

Other Notes

To view sequence information for this product, please visit the product page


Storage Class

12 - Non Combustible Liquids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library