Skip to Content
MilliporeSigma

OGS1314

PSF-CMV-PURO-COOH-THR-RLUC-6HIS - C-TERMINAL 6 HIS AND RLUC DUAL TAG MAMMALIAN PLASMID

plasmid vector for molecular cloning

Synonym(s):

cloning vector, expression vector, molecular cloning vector, plasmid, plasmid vector, snapfast vector, vector

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.85
UNSPSC Code:
12352200
eCl@ss:
32160703
Promoter:
Promoter name: CMV
Promoter activity: constitutive
Promoter type: mammalian
Origin of replication:
pUC (500 copies)
Bacteria selection:
kanamycin
Reporter gene:
Renilla luciferase
Peptide cleavage:
thrombin
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


conjugate

Renilla luciferase conjugate

tag

6-His tagged

form

buffered aqueous solution

mol wt

size 7119 bp

technique(s)

molecular cloning: suitable

bacteria selection

kanamycin

mammalian cells selection

puromycin

origin of replication

pUC (500 copies)

peptide cleavage

thrombin

peptide tag location

C-terminal

promoter

Promoter name: CMV
Promoter activity: constitutive
Promoter type: mammalian

reporter gene

Renilla luciferase

shipped in

ambient

storage temp.

−20°C

General description

This plasmid is designed to express tagged proteins in mammalian cells either by transient transfection or by creating stable cell lines. It contains a puromycin resistance expression cassette using the human Ubiquitin promoter to drive expression and allow for the selection of cells containing the plasmid.

About the Peptide Tag:This plasmid contains a c-terminal Renilla luciferase reporter tag that can be fused to a gene of interest to allow protein detection. This plasmid also contains a secondary Hexa-Histidine (6His) tag protein tag. The sequence of this tag is: HHHHHHWe provide a range of dual peptide tag plasmids. This is because some peptide tags provide specific biological properties (e. g., small molecule affinity new epitopes solubility or protein secretion) that are not provided by others.

About the Cleavage Tag: This plasmid also encodes a protease cleavage site that is designed to be positioned between your gene of interest and the tag to allow the removal of the tag following protein purification or isolation. This plasmid contains a Thrombin cleavage tag. The protein sequence of the cleavage tag is: LVPRGS. It cleaves preferentially between the Arg and Gly residues. Off target cleavage can often occur at non-specific sites normally from other contaminating proteases. To ensure maximal protein integrity the enzyme reagent must be highly pure.

Promoter Expression Level: This plasmid contains the mammalian CMV promoter to drive gene expression. We have tested all of our mammalian promoters in a range of cell types and CMV is consistently the strongest in those we have studied. However there are many reports of the CMV promoter demonstrating silencing by methylation in long-term culture.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.oxfordgenetics.com.

Other Notes

Looking for more vector options to move your experiments forward faster? Consider a custom cloning vector designed and built by Oxford Genetics™. Find out more at Oxford Genetics - Sigma's partner for cloning and expression vectors for molecular biology and synthetic biology applications.
Please select the file type you require. For reference most cloning programs will import a .gb (Genbank) file and will show all of the plasmids features automatically when downloaded and imported.

Genebank Vector Sequence File

FASTA Vector Sequence File

Full Plasmid Map

Legal Information

Oxford Genetics is a trademark of Oxford Genetics Ltd


Storage Class

12 - Non Combustible Liquids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library