Skip to Content
MilliporeSigma

OGS3025

PSF-OXB20-COOH-3C-BETAGAL-6HIS - C-TERMINAL 6 HIS AND BETA GAL DUAL TAG BACTERIAL PLASMID

plasmid vector for molecular cloning

Synonym(s):

cloning vector, expression vector, molecular cloning vector, plasmid, plasmid vector, snapfast vector, vector

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.85
UNSPSC Code:
12352200
Promoter:
Promoter name: OXB20
Promoter activity: constitutive
Promoter type: bacterial
Origin of replication:
pUC (500 copies)
Bacteria selection:
kanamycin
Reporter gene:
beta Gal
Peptide cleavage:
3C
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


recombinant

expressed in E. coli

tag

6-His tagged, beta-galactosidase tagged

form

buffered aqueous solution

mol wt

size 7037 bp

technique(s)

molecular cloning: suitable

bacteria selection

kanamycin

origin of replication

pUC (500 copies)

peptide cleavage

3C

peptide tag location

C-terminal

promoter

Promoter name: OXB20
Promoter activity: constitutive
Promoter type: bacterial

reporter gene

beta Gal

shipped in

ambient

storage temp.

−20°C

General description

This plasmid is designed to express tagged proteins in E. coli. The plasmid contains a constitutive promoter (OXB20) derived from the region upstream of the E. coli RecA gene. It does not require induction or any additional components for activity. It is the strongest of the bacterial promoters that we provide and this high level of expression can cause expression problems with some proteins with poor solubility. For this reason we sell a range of bacterial promoters with different expression levels (OXB1(low)>OXB20(high)) that can be provided with the peptide tags in this plasmid on request.

About the Peptide Tag:This plasmid contains a c-terminal Beta Galactosidase reporter tag that can be fused to a gene of interest to allow protein detection. This plasmid also contains a secondary Hexa-Histidine (6His) tag protein tag. The sequence of this tag is: HHHHHHWe provide a range of dual peptide tag plasmids. This is because some peptide tags provide specific biological properties (e. g., small molecule affinity new epitopes solubility or protein secretion) that are not provided by others.

About the Cleavage Tag: This plasmid also encodes a protease cleavage site that is designed to be positioned between your gene of interest and the tag to allow the removal of the tag following protein purification or isolation. This plasmid contains a 3C cleavage tag. The protein sequence of the cleavage tag is: LEVLFQGP. Human Rhinovirus (HRV) 3C Protease is a highly specific protease that cleaves between the Glu and Gly residues of its recognition site. It is often produced with the trademname ′PreScission protease′.

Promoter Expression Level: This plasmid contains a constitutive bacterial promoter that does not require induction. It is the strongest bacterial promoter we sell and this can cause solubility and expression problems with some proteins. We also offer a range of other bacterial promoters that are compatible with this plasmid and are available on request.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.oxgene.com

Other Notes

To view sequence information for this product, please visit the product page


Storage Class

12 - Non Combustible Liquids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library